2-Bromoisophthalaldehyde, 97%, Thermo Scientific Chemicals
2-Bromoisophthalaldehyde, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Bromoisophthalaldehyde, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number H64423.06
also known as H64423-06
Price (USD)/ Each
219.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
219.00
Add to cart
2-Bromoisophthalaldehyde, 97%, Thermo Scientific Chemicals
Catalog numberH64423.06
Price (USD)/ Each
219.00
-
Add to cart
Chemical Identifiers
CAS79839-49-9
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>96.0%
Used in the preparation of paclitaxel-mimicking alkaloids with anticancer activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in the preparation of paclitaxel-mimicking alkaloids with anticancer activity.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. Yong-Kang Zhang; Jacob J. Plattner; Eric E. Easom; David Waterson; Min Gec; Zhiya Lic; Lingchao Lic; Yong Jian. An efficient synthesis for a new class antimalarial agent, 7-(2-carboxyethyl)-1,3-dihydro-1-hydroxy-2,1-benzoxaborole. Tetrahedron Letters. 201152 (30), 3909-3911.
  2. Jin-Hua Zhang; Ping Li; Wen-Ping Hub; Hong-Xing Wang. Substituent effect of diimino-palladium (II) pincer complexes on the catalysis of Sonogashira coupling reaction. Polyhedron. 2011 (96), 107-112.