Used in the preparation of α-aryl/pyridinyl ethanolamines as selective β3 adrenergic receptor agonists.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Used in the preparation of α-aryl/pyridinyl ethanolamines as selective β3 adrenergic receptor agonists.
Solubility
Soluble in DMSO and chloroform. Slightly soluble in water.
Notes
Store away from acids and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only
J. Trögl, et al. Automatic formation of hypotheses on the relationships between structure of naphthalene analogs and bioluminescence response of bioreporter Pseudomonas fluorescens HK44.Folia Microbiologica.,2010,555(5), 411-417.
B. Lovelock, et al. Properties of 4-octadecycloxy-1-naphthoic acid in micellar solutions and in monolayer films absorbed onto silica attenuated total reflectance plates.J. Phys. Chem.,1985,89(3), 501-507.