Used as a intermediate for organic light-emitting diode(OLED) and pharmaceutical.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Used as a intermediate for organic light-emitting diode(OLED) and pharmaceutical.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
RUO – Research Use Only
General References:
- Kunio Saito; and Yasuo Kikugawa. Introduction of a methoxyl group into indoles and 2,3-dihydroindoles. A new synthesis of 5-hydroxytryptamine (serotonin). Journal of Heterocyclic Chemistry. 197916 (7), 1325-1328.
- Holger Spanggaard; Mikkel Jørgensen; and Kristoffer Almdal. Mechanical Strain Sensing in a SIS-Type Elastomer with Single Site Strain Probes Based on Carbazole. Macromolecules. 200336 (5), 1701-1705.