Thermo Scientific Chemicals

3,6-Dihydro-2H-pyran-4-boronic acid pinacol ester, 98%, Thermo Scientific Chemicals

Catalog number: H64452.06
5 g, Each
Thermo Scientific Chemicals

3,6-Dihydro-2H-pyran-4-boronic acid pinacol ester, 98%, Thermo Scientific Chemicals

Catalog number: H64452.06
5 g, Each
Quantity
Catalog number: H64452.06
also known as H64452-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
287944-16-5
IUPAC Name
2-(3,6-dihydro-2H-pyran-4-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Formula
C11H19BO3
InChI Key
DOSGEBYQRMBTGS-UHFFFAOYSA-N
SMILES
CC1(C)OB(OC1(C)C)C1=CCOCC1
Assay (HPLC)
>97.5%

Description

It plays an important role as intermediate for organic synthesis agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It plays an important role as intermediate for organic synthesis agrochemical, pharmaceutical and dyestuff field.

Solubility
Slightly soluble in water.

Notes
Moisture Sensitive. Store away form water/moisture. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text