6-Azaindole is an indole derivative and useful synthetic intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
6-Azaindole is an indole derivative and useful synthetic intermediate.
Solubility
Soluble in water (hardly), methanol and chloroform.
Notes
Air and Light Sensitive. Protect from light store in dark. Store away from air.
RUO – Research Use Only
General References:
- Susan M. Twine; Lise Murphy; Robert S. Phillips; Patrik Callis; Michael T. Cash; and Arthur G. Szabo . The Photophysical Properties of 6-Azaindole. J. Phys. Chem. B. 2003 107(2), 637-645.
- Zhongxing Zhang; Zhong Yang; Nicholas A. Meanwell; John F. Kadow; and Tao Wang. A General Method for the Preparation of 4- and 6-Azaindoles. J. Org. Chem. 2002 67(7), 2345-2347.