Thermo Scientific Chemicals

1-Benzyloxycarbonyl-3-piperidone, 97+%, Thermo Scientific Chemicals

Catalog number: H64471.14
25 g, Each
Thermo Scientific Chemicals

1-Benzyloxycarbonyl-3-piperidone, 97+%, Thermo Scientific Chemicals

Catalog number: H64471.14
25 g, Each
Quantity
Catalog number: H64471.14
also known as H64471-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
61995-20-8
Assay (GC)
>97.0%

Description

1-Benzyloxycarbonyl-3-piperidone finds its application in the synthesis and pharmacological evaluation of some 6-substituted 7-methyl-1,4-dioxa-7-azaspiro[4.5]decanes as potential dopamine agonists.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Benzyloxycarbonyl-3-piperidone finds its application in the synthesis and pharmacological evaluation of some 6-substituted 7-methyl-1,4-dioxa-7-azaspiro[4.5]decanes as potential dopamine agonists.

Solubility
Slightly soluble in water.

Notes
Store away from heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep it refrigerated.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text