1,5-Naphthyridine, 97+%, Thermo Scientific Chemicals
1,5-Naphthyridine, 97+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,5-Naphthyridine, 97+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Catalog number H64484.MD
also known as H64484-MD
Price (USD)/ Each
59.40
-
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (USD)/ Each
59.40
Add to cart
1,5-Naphthyridine, 97+%, Thermo Scientific Chemicals
Catalog numberH64484.MD
Price (USD)/ Each
59.40
-
Add to cart
Chemical Identifiers
CAS254-79-5
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only