2-[2-(Trifluoromethyl)phenyl]ethylamine, 95%, Thermo Scientific Chemicals
2-[2-(Trifluoromethyl)phenyl]ethylamine, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-[2-(Trifluoromethyl)phenyl]ethylamine, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number H64492.03
also known as H64492-03
Price (USD)/ Each
56.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
56.00
Add to cart
2-[2-(Trifluoromethyl)phenyl]ethylamine, 95%, Thermo Scientific Chemicals
Catalog numberH64492.03
Price (USD)/ Each
56.00
-
Add to cart
Chemical Identifiers
CAS774-18-5
IUPAC Name2-[2-(trifluoromethyl)phenyl]ethan-1-amine
Molecular FormulaC9H10F3N
InChI KeyLIERORLYMWHXDL-UHFFFAOYSA-N
SMILESNCCC1=CC=CC=C1C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
Assay (GC)>94.0%
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only