Thermo Scientific Chemicals

2-(3-Methoxyphenyl)ethylamine, 97+%, Thermo Scientific Chemicals

Catalog number: H64494.06
5 g, Each
Thermo Scientific Chemicals

2-(3-Methoxyphenyl)ethylamine, 97+%, Thermo Scientific Chemicals

Catalog number: H64494.06
5 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: H64494.06
also known as H64494-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2039-67-0
IUPAC Name
hydrogen 2-(3-methoxyphenyl)ethan-1-amine chloride
Molecular Formula
C9H14ClNO
InChI Key
KDRBQDYHPKRFGX-UHFFFAOYSA-N
SMILES
[H+].[Cl-].COC1=CC=CC(CCN)=C1
Assay (GC)
>97.0%

Description

Activated phenethylamine used in the perfluorooctanesulfonic acid catalyzed Pictet-Spengler reaction. Also used in a synthesis of 1,3-oxazepines via palladium-catalyzed intramolecular coupling.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Activated phenethylamine used in the perfluorooctanesulfonic acid catalyzed Pictet-Spengler reaction. Also used in a synthesis of 1,3-oxazepines via palladium-catalyzed intramolecular coupling.

Solubility
Insoluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text