2-(3-Methoxyphenyl)ethylamine, 97+%, Thermo Scientific Chemicals
2-(3-Methoxyphenyl)ethylamine, 97+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-(3-Methoxyphenyl)ethylamine, 97+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number H64494.14
also known as H64494-14
Price (USD)/ Each
86.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
102.00 
Save 15.35 (15%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
86.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
102.00 
Save 15.35 (15%)
Add to cart
2-(3-Methoxyphenyl)ethylamine, 97+%, Thermo Scientific Chemicals
Catalog numberH64494.14
Price (USD)/ Each
86.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
102.00 
Save 15.35 (15%)
-
Add to cart
Chemical Identifiers
CAS2039-67-0
IUPAC Namehydrogen 2-(3-methoxyphenyl)ethan-1-amine chloride
Molecular FormulaC9H14ClNO
InChI KeyKDRBQDYHPKRFGX-UHFFFAOYSA-N
SMILES[H+].[Cl-].COC1=CC=CC(CCN)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)>97.0%
Activated phenethylamine used in the perfluorooctanesulfonic acid catalyzed Pictet-Spengler reaction. Also used in a synthesis of 1,3-oxazepines via palladium-catalyzed intramolecular coupling.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Activated phenethylamine used in the perfluorooctanesulfonic acid catalyzed Pictet-Spengler reaction. Also used in a synthesis of 1,3-oxazepines via palladium-catalyzed intramolecular coupling.

Solubility
Insoluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. G P Miller; I H Hanna; Y Nishimura; F P Guengerich. Oxidation of phenethylamine derivatives by cytochrome P450 2D6: the issue of substrate protonation in binding and catalysis. J. Comb. Chem. 2001 40(47), 14215-14223.
  2. P T Wong; W L Teo; T Lee. A microdialysis study on striatal dopamine, 5-HT and metabolites in conscious rats after various treatments: evidence for extravesicular release of dopamine. Neuroreport. 1993 5(1), 53-56.