Chemical Identifiers
CAS123847-85-8
IUPAC NameN4,N4'-bis(naphthalen-1-yl)-N4,N4'-diphenyl-[1,1'-biphenyl]-4,4'-diamine
Molecular FormulaC44H32N2
InChI KeyIBHBKWKFFTZAHE-UHFFFAOYSA-N
SMILESC1=CC=C(C=C1)N(C1=CC=C(C=C1)C1=CC=C(C=C1)N(C1=CC=CC=C1)C1=C2C=CC=CC2=CC=C1)C1=C2C=CC=CC2=CC=C1
View more