An aldehyde containing building block used for proteomics research
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
An aldehyde containing building block used for proteomics research
Solubility
Slightly soluble in water.
Notes
Air Sensitive. Store away form air. Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
RUO – Research Use Only
Hing L. Sham; David A. Betebenner; Xiaoqi Chen; Ayda Saldivar; Sudthida Vasavanonda; Dale J. Kempf; Jacob J. Plattner; Daniel W. Norbeck. Synthesis and structure-activity relationships of a novel series of HIV-1 protease inhibitors encompassing ABT-378 (Lopinavir). Bioorganic & Medicinal Chemistry Letters. 1972 12(8), 1185-1187.
C. L. Bell; S. S. Danyluk; T. Schaefer.Signs of spin-spin coupling constants between aldehydic and ring protons in 2,6-dinitrobenzaldehyde. Evidence for a hyperconjugative contribution to JpH,CHO. Canadian Journal of Chemistry. 1969 47(19), 3529-3533.