Thermo Scientific Chemicals

1-Bromo-3,4,5-trichlorobenzene, 98%, Thermo Scientific Chemicals

Catalog number: H64506.14
25 g, Each
Thermo Scientific Chemicals

1-Bromo-3,4,5-trichlorobenzene, 98%, Thermo Scientific Chemicals

Catalog number: H64506.14
25 g, Each
Quantity
Catalog number: H64506.14
also known as H64506-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
21928-51-8
IUPAC Name
5-bromo-1,2,3-trichlorobenzene
Molecular Formula
C6H2BrCl3
InChI Key
VZUMVBQMJFFYRM-UHFFFAOYSA-N
SMILES
ClC1=CC(Br)=CC(Cl)=C1Cl
Assay (HPLC)
>97.5%

Description

It is a multipurpose intermediate commonly used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a multipurpose intermediate commonly used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text