Specifications
Chemical Name or Material6-Hydroxy-1,2,3,4-tetrahydroisoquinoline
CAS14446-24-3
Health Hazard 1H315-H319-H335
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
Melting Point192°C to 193°C
View more