Thermo Scientific Chemicals

Benzenesulfonic acid isopropyl ester, 95%, Thermo Scientific Chemicals

Catalog number: H64523.06
5 g, Each
Thermo Scientific Chemicals

Benzenesulfonic acid isopropyl ester, 95%, Thermo Scientific Chemicals

Catalog number: H64523.06
5 g, Each
Quantity
Catalog number: H64523.06
also known as H64523-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6214-18-2
IUPAC Name
2-methylpropyl benzenesulfonate
Molecular Formula
C10H14O3S
InChI Key
NFRRFEFZKGNYKS-UHFFFAOYSA-N
SMILES
CC(C)COS(=O)(=O)C1=CC=CC=C1
Assay (HPLC)
>94.0%

Description

A sulfonate ester that acts as a potential effects in drug substances impurity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A sulfonate ester that acts as a potential effects in drug substances impurity.

Solubility
Soluble in chloroform and ethyl acetate.

Notes
Store at 4°C. Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text