2,4-Dichloro-6-nitrophenol, 98+%, Thermo Scientific Chemicals
2,4-Dichloro-6-nitrophenol, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,4-Dichloro-6-nitrophenol, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number H64524.22
also known as H64524-22
Price (USD)/ Each
296.00
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
296.00
Add to cart
2,4-Dichloro-6-nitrophenol, 98+%, Thermo Scientific Chemicals
Catalog numberH64524.22
Price (USD)/ Each
296.00
-
Add to cart
Chemical Identifiers
CAS609-89-2
IUPAC Name2,4-dichloro-6-nitrophenol
Molecular FormulaC6H3Cl2NO3
InChI KeyLYPMXMBQPXPNIQ-UHFFFAOYSA-N
SMILESOC1=C(Cl)C=C(Cl)C=C1[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>98.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only