Thermo Scientific Chemicals

3-(Aminomethyl)benzoic acid hydrochloride, 95%, Thermo Scientific Chemicals

Catalog number: H64546.03
1 g, Each
Thermo Scientific Chemicals

3-(Aminomethyl)benzoic acid hydrochloride, 95%, Thermo Scientific Chemicals

Catalog number: H64546.03
1 g, Each
Quantity
Catalog number: H64546.03
also known as H64546-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
876-03-9
IUPAC Name
3-(aminomethyl)benzoic acid hydrochloride
Molecular Formula
C8H10ClNO2
InChI Key
SJCCOASSOPUHEN-UHFFFAOYSA-N
SMILES
Cl.NCC1=CC=CC(=C1)C(O)=O
Assay (Titration)
>94.0%

Description

It is used as pharmaceutical intermediate. Derivatives of benzylamine and benzamidine were found to be competitive inhibitors of the proteolytic enzymes trypsin, plasmin, and thrombin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediate. Derivatives of benzylamine and benzamidine were found to be competitive inhibitors of the proteolytic enzymes trypsin, plasmin, and thrombin.

Solubility
Slightly soluble in water.

Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text