Thermo Scientific Chemicals

4-Pyridineacetonitrile, ≥97%, Thermo Scientific Chemicals

Catalog number: H64588.03
1 g, Each
Thermo Scientific Chemicals

4-Pyridineacetonitrile, ≥97%, Thermo Scientific Chemicals

Catalog number: H64588.03
1 g, Each
Quantity
Catalog number: H64588.03
also known as H64588-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13121-99-8
IUPAC Name
2-(pyridin-4-yl)acetonitrile
Molecular Formula
C7H6N2
InChI Key
BMVSAKPRNWZCPG-UHFFFAOYSA-N
SMILES
N#CCC1=CC=NC=C1
Assay (HPLC)
>97.0%

Description

A pyridine derivative used in the preparation of inhibitors of Acyl CoA, cholesterol acyltransferase as well as bone morphogenetic protein (BMP) signaling inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A pyridine derivative used in the preparation of inhibitors of Acyl CoA, cholesterol acyltransferase as well as bone morphogenetic protein (BMP) signaling inhibitors.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text