3-Fluoro-4-methoxypyridine, 98%, Thermo Scientific Chemicals
3-Fluoro-4-methoxypyridine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Fluoro-4-methoxypyridine, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number H64603.06
also known as H64603-06
Price (USD)/ Each
436.65
Online exclusive
485.00 
Save 48.35 (10%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
436.65
Online exclusive
485.00 
Save 48.35 (10%)
Add to cart
3-Fluoro-4-methoxypyridine, 98%, Thermo Scientific Chemicals
Catalog numberH64603.06
Price (USD)/ Each
436.65
Online exclusive
485.00 
Save 48.35 (10%)
-
Add to cart
Chemical Identifiers
CAS1060805-03-9
IUPAC Name3-fluoro-4-methoxypyridine
Molecular FormulaC6H6FNO
InChI KeyXYTUUTJNWFECNS-UHFFFAOYSA-N
SMILESCOC1=C(F)C=NC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear colorless to pale yellow
Assay from Supplier's CofA≥96.0% (HPLC)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only