2-Bromo-2'-methylacetophenone, 98%, Thermo Scientific Chemicals
2-Bromo-2'-methylacetophenone, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Bromo-2'-methylacetophenone, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number H64651.06
also known as H64651-06
Price (USD)/ Each
350.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
350.00
Add to cart
2-Bromo-2'-methylacetophenone, 98%, Thermo Scientific Chemicals
Catalog numberH64651.06
Price (USD)/ Each
350.00
-
Add to cart
Chemical Identifiers
CAS51012-65-8
IUPAC Name2-bromo-1-(2-methylphenyl)ethan-1-one
Molecular FormulaC9H9BrO
InChI KeyXMGAXELQRATLJP-UHFFFAOYSA-N
SMILESCC1=CC=CC=C1C(=O)CBr
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%
2-Bromo-2'-methylacetophenone is an aromatic building block for proteomics research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-2′-methylacetophenone is an aromatic building block for proteomics research.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. Matthias Nettekoven; Wolfgang Guba; Werner Neidhart; Patrizio Mattei; Philippe Pflieger; Olivier Roche; Sven Taylor. Isomeric thiazole derivatives as ligands for the neuropeptide Y5 receptor. Bioorganic & Medicinal Chemistry Letters. 2005 15(14), 3446-3449.
  2. Matthias Nettekoven Dr.; Wolfgang Guba Dr.; Werner Neidhart Dr.; Patrizio Mattei Dr.; Philippe Pflieger Dr.; Jean-Marc Plancher Dr.; and Sven Taylor Dr. Aminothiazole Derivatives as Neuropeptide Y5 Receptor Ligands: Finding the Balance between Affinity and Physicochemical Properties. ChemMedChem. 2006 1(1), 45-48.