Thioisobutyramide, 95%, Thermo Scientific Chemicals
Thioisobutyramide, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Thioisobutyramide, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Catalog number H64663.14
also known as H64663-14
Price (USD)/ Each
636.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
636.00
Add to cart
Thioisobutyramide, 95%, Thermo Scientific Chemicals
Catalog numberH64663.14
Price (USD)/ Each
636.00
-
Add to cart
Chemical Identifiers
CAS13515-65-6
IUPAC Name2-methylpropanethioamide
Molecular FormulaC4H9NS
InChI KeyNPCLRBQYESMUPD-UHFFFAOYSA-N
SMILESCC(C)C(N)=S
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>94.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only