Thermo Scientific Chemicals

Naphthalene-2-boronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Catalog number: H64671.06
5 g, Each
Thermo Scientific Chemicals

Naphthalene-2-boronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Catalog number: H64671.06
5 g, Each
Quantity
Catalog number: H64671.06
also known as H64671-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
256652-04-7
IUPAC Name
4,4,5,5-tetramethyl-2-(naphthalen-2-yl)-1,3,2-dioxaborolane
Molecular Formula
C16H19BO2
InChI Key
SPPZBAGKKBHZRW-UHFFFAOYSA-N
SMILES
CC1(C)OB(OC1(C)C)C1=CC=C2C=CC=CC2=C1
Assay (HPLC)
>96.0%

Description

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Slightly soluble in water.

Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text