3-Bromobenzamide, 98+%, Thermo Scientific Chemicals
3-Bromobenzamide, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Bromobenzamide, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number H64672.22
also known as H64672-22
Price (USD)/ Each
202.00
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
202.00
Add to cart
3-Bromobenzamide, 98+%, Thermo Scientific Chemicals
Catalog numberH64672.22
Price (USD)/ Each
202.00
-
Add to cart
Chemical Identifiers
CAS22726-00-7
IUPAC Name3-bromobenzamide
Molecular FormulaC7H6BrNO
InChI KeyODJFDWIECLJWSR-UHFFFAOYSA-N
SMILESNC(=O)C1=CC=CC(Br)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)≥98.0%
Appearance (Color)White to off-white
FormPowder or crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only