Thermo Scientific Chemicals

4-Bromo-1H-pyrazole, ≥98%, Thermo Scientific Chemicals

Catalog number: H64673.14
25 g, Each
Thermo Scientific Chemicals

4-Bromo-1H-pyrazole, ≥98%, Thermo Scientific Chemicals

Catalog number: H64673.14
25 g, Each
Quantity
Catalog number: H64673.14
also known as H64673-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2075-45-8
IUPAC Name
4-bromo-1H-pyrazole
Molecular Formula
C3H3BrN2
InChI Key
WVGCPEDBFHEHEZ-UHFFFAOYSA-N
SMILES
BrC1=CNN=C1
Assay (HPLC)
>98.0%

Description

4-Bromopyrazole may be used in the preparation of 4-bromo-1-(2-chloroethyl)-1H-pyrazole. It may be used as starting material in the synthesis of 1,4?-bipyrazoles. Also used in the synthesis of various pharmaceutical and biologically active compounds, including inhibitors. It is also shown to be one of the 4-substituted pyrazoles, being able to act as inhibitor of liver alcohol dehydrogenase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromopyrazole may be used in the preparation of 4-bromo-1-(2-chloroethyl)-1H-pyrazole. It may be used as starting material in the synthesis of 1,4′-bipyrazoles. Also used in the synthesis of various pharmaceutical and biologically active compounds, including inhibitors. It is also shown to be one of the 4-substituted pyrazoles, being able to act as inhibitor of liver alcohol dehydrogenase.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text