Thermo Scientific Chemicals

2-Amino-4-chloro-3-nitropyridine, 95%, Thermo Scientific Chemicals

Catalog number: H64678.06
5 g, Each
Thermo Scientific Chemicals

2-Amino-4-chloro-3-nitropyridine, 95%, Thermo Scientific Chemicals

Catalog number: H64678.06
5 g, Each
Quantity
Catalog number: H64678.06
also known as H64678-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6980-08-1
IUPAC Name
4-chloro-3-nitropyridin-2-amine
Molecular Formula
C5H4ClN3O2
InChI Key
DIRINUVNYFAWQF-UHFFFAOYSA-N
SMILES
NC1=NC=CC(Cl)=C1[N+]([O-])=O
Assay (HPLC)
>94.0%

Description

2-Amino-4-chloro-3-nitropyridine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-4-chloro-3-nitropyridine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Soluble in dimethyl formamide, dimethyl sulfoxide, hot ethanol, ethyl acetate and hot methanol.

Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text