4(1H)-Pyrimidinone, 98%, Thermo Scientific Chemicals
4(1H)-Pyrimidinone, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4(1H)-Pyrimidinone, 98%, Thermo Scientific Chemicals

Have Questions?
Quantity:
5 g
Catalog number H64690.06
also known as H64690-06
Price (USD)/ Each
97.70
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
97.70
Add to cart
4(1H)-Pyrimidinone, 98%, Thermo Scientific Chemicals
Catalog numberH64690.06
Price (USD)/ Each
97.70
-
Add to cart
Chemical Identifiers
CAS4562-27-0
IUPAC Name3,4-dihydropyrimidin-4-one
Molecular FormulaC4H4N2O
InChI KeyDNCYBUMDUBHIJZ-UHFFFAOYSA-N
SMILESO=C1NC=NC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder or crystals or crystalline powder
Appearance (Color)White to pale yellow
Assay from Supplier's CofA≥97.5% (GC)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only