2-Amino-5-fluoro-6-methylpyridine, 98%, Thermo Scientific Chemicals
2-Amino-5-fluoro-6-methylpyridine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Amino-5-fluoro-6-methylpyridine, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H64700.03
also known as H64700-03
Price (USD)/ Each
88.50
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
88.50
Add to cart
2-Amino-5-fluoro-6-methylpyridine, 98%, Thermo Scientific Chemicals
Catalog numberH64700.03
Price (USD)/ Each
88.50
-
Add to cart
Chemical Identifiers
CAS110919-71-6
IUPAC Name5-fluoro-6-methylpyridin-2-amine
Molecular FormulaC6H7FN2
InChI KeyWMGRXNKQEVOVET-UHFFFAOYSA-N
SMILESCC1=C(F)C=CC(N)=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only