3-Fluoro-2-methylpyridine was used in the synthesis of fluorine-containing pyridinealdoximes of potential use for the reactivation of cholinesterase.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Fluoro-2-methylpyridine was used in the synthesis of fluorine-containing pyridinealdoximes of potential use for the reactivation of cholinesterase.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
RUO – Research Use Only
Kensuke Kobayashi; Tomohiro Tsujita; Hirokatsu Ito; Satoshi Ozaki; Takeshi Tani; Yasuyuki Ishii; Shoki Okuda; Kiyoshi Tadano; Takahiro Fukuroda; Hisashi Ohta; Osamu Okamoto. Identification of MK-1925: A selective, orally active and brain-penetrable opioid receptor-like 1 (ORL1) antagonist. Bioorganic & Medicinal Chemistry Letters. 2009 19(16), 4729-4732.
Karikere Ekanna Manojkumar; Swamy Sreenivasa; Mohan Revanasiddappa Nadigar; and Tadimety Harikrishna .6-[5-Chloro-2-(trifluoromethyl)phenyl]-3-fluoro-2-methylpyridine. Molbank. 2013 4