Methyl 4-chloroacetoacetate, 97+%, Thermo Scientific Chemicals
Methyl 4-chloroacetoacetate, 97+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methyl 4-chloroacetoacetate, 97+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number H64709.22
also known as H64709-22
Price (USD)/ Each
137.00
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
137.00
Add to cart
Methyl 4-chloroacetoacetate, 97+%, Thermo Scientific Chemicals
Catalog numberH64709.22
Price (USD)/ Each
137.00
-
Add to cart
Chemical Identifiers
CAS32807-28-6
IUPAC Namemethyl 4-chloro-3-oxobutanoate
Molecular FormulaC5H7ClO3
InChI KeyHFLMYYLFSNEOOT-UHFFFAOYSA-N
SMILESCOC(=O)CC(=O)CCl
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only