Thermo Scientific Chemicals

2-Bromo-9,9-dimethylfluorene, 98%, Thermo Scientific Chemicals

Catalog number: H64732.14
25 g, Each
Thermo Scientific Chemicals

2-Bromo-9,9-dimethylfluorene, 98%, Thermo Scientific Chemicals

Catalog number: H64732.14
25 g, Each
Quantity
Catalog number: H64732.14
also known as H64732-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
28320-31-2
IUPAC Name
2-bromo-9,9-dimethyl-9H-fluorene
Molecular Formula
C15H13Br
InChI Key
MBHPOBSZPYEADG-UHFFFAOYSA-N
SMILES
CC1(C)C2=CC=CC=C2C2=C1C=C(Br)C=C2
Assay (GC)
98%

Description

This material is a popular synthetic precursor for OLED materials. Also used to make devices that efficiently emit a deep blue color.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
This material is a popular synthetic precursor for OLED materials. Also used to make devices that efficiently emit a deep blue color.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text