Methyl 5-bromo-6-chloronicotinate, 98%, Thermo Scientific Chemicals
Methyl 5-bromo-6-chloronicotinate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methyl 5-bromo-6-chloronicotinate, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H64754.06
also known as H64754-06
Price (USD)/ Each
85.10
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
85.10
Add to cart
Methyl 5-bromo-6-chloronicotinate, 98%, Thermo Scientific Chemicals
Catalog numberH64754.06
Price (USD)/ Each
85.10
-
Add to cart
Chemical Identifiers
CAS78686-77-8
IUPAC Namemethyl 5-bromo-6-chloropyridine-3-carboxylate
Molecular FormulaC7H5BrClNO2
InChI KeyWINGWVOUOFMOJQ-UHFFFAOYSA-N
SMILESCOC(=O)C1=CN=C(Cl)C(Br)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only