Used in organic synthesis, catalytic agent, petrochemical additive and synthetic chemistry. Also used as intermediate for pharmaceutical.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Used in organic synthesis, catalytic agent, petrochemical additive and synthetic chemistry. Also used as intermediate for pharmaceutical.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
RUO – Research Use Only
DJ Brown; and T Lee. Pyrimidine reactions. XVI. Thermal rearrangement of substituted 2- and 4-alkoxypyrimidines. Australian Journal of Chemistry.1968 21(1), 243 - 255.
Lucjan Strekowski; Donald B. Harden; William B. Grubb III; Steven E. Patterson; Agnieszka Czarny; Maria J. Mokrosz; Marek T. Cegla; and Roman L. Wydra. Synthesis of 2-chloro-4,6-di(heteroaryl)pyrimidines. Journal of Heterocyclic Chemistry.1990 27(5),1393-1400.