Thermo Scientific Chemicals

Ethyl 4-formylpyrrole-2-carboxylate, 96%, Thermo Scientific Chemicals

Catalog number: H64776.03
1 g, Each
Thermo Scientific Chemicals

Ethyl 4-formylpyrrole-2-carboxylate, 96%, Thermo Scientific Chemicals

Catalog number: H64776.03
1 g, Each
Quantity
Catalog number: H64776.03
also known as H64776-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7126-57-0
IUPAC Name
ethyl 4-formyl-1H-pyrrole-2-carboxylate
Molecular Formula
C8H9NO3
InChI Key
WVSAWXIWWNJTAV-UHFFFAOYSA-N
SMILES
CCOC(=O)C1=CC(C=O)=CN1
Assay (HPLC)
>95.0%

Description

Ethyl 4-formylpyrrole-2-carboxylate is a pyrrole building block for proteomics research. Further, it is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 4-formylpyrrole-2-carboxylate is a pyrrole building block for proteomics research. Further, it is used as a pharmaceutical intermediate.

Solubility
Miscible with ethyl acetate.

Notes
Incompatible with steel and aluminum.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text