3-Bromo-5-nitrobenzaldehyde, ≥97%, Thermo Scientific Chemicals
3-Bromo-5-nitrobenzaldehyde, ≥97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Bromo-5-nitrobenzaldehyde, ≥97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H64787.03
also known as H64787-03
Price (USD)/ Each
123.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
123.00
Add to cart
3-Bromo-5-nitrobenzaldehyde, ≥97%, Thermo Scientific Chemicals
Catalog numberH64787.03
Price (USD)/ Each
123.00
-
Add to cart
Chemical Identifiers
CAS355134-13-3
IUPAC Name3-bromo-5-nitrobenzaldehyde
Molecular FormulaC7H4BrNO3
InChI KeyOUDCOMBHRXKPIJ-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC(Br)=CC(C=O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>= 97.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only