Thermo Scientific Chemicals

Ethyl 2,4-dichloropyrimidine-5-carboxylate, 98%, Thermo Scientific Chemicals

Catalog number: H64788.03
1 g, Each
Thermo Scientific Chemicals

Ethyl 2,4-dichloropyrimidine-5-carboxylate, 98%, Thermo Scientific Chemicals

Catalog number: H64788.03
1 g, Each
Quantity
Catalog number: H64788.03
also known as H64788-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
51940-64-8
IUPAC Name
ethyl 2,4-dichloropyrimidine-5-carboxylate
Molecular Formula
C7H6Cl2N2O2
InChI Key
SRJBDGLSCPDXBL-UHFFFAOYSA-N
SMILES
CCOC(=O)C1=CN=C(Cl)N=C1Cl
Assay (HPLC)
>97.5%

Description

It's employed as an important intermediate for raw material for organic synthesis, agrochemical, pharmaceutical and dyestuff field

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It′s employed as an important intermediate for raw material for organic synthesis, agrochemical, pharmaceutical and dyestuff field

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text