Thermo Scientific Chemicals

9-Bromo-1-nonene, 97%, Thermo Scientific Chemicals

Catalog number: H64792.06
5 g, Each
Thermo Scientific Chemicals

9-Bromo-1-nonene, 97%, Thermo Scientific Chemicals

Catalog number: H64792.06
5 g, Each
Quantity
Catalog number: H64792.06
also known as H64792-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
89359-54-6
IUPAC Name
9-bromonon-1-ene
Molecular Formula
C9H17Br
InChI Key
RQXPBVHYVAOUBY-UHFFFAOYSA-N
SMILES
BrCCCCCCCC=C
Assay (HPLC)
>96.0%

Description

It's employed as an important intermediate for raw material for organic synthesis, agrochemical, pharmaceutical and dyestuff field

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It′s employed as an important intermediate for raw material for organic synthesis, agrochemical, pharmaceutical and dyestuff field

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text