Methyl 4-methylsalicylate, 98+%, Thermo Scientific Chemicals
Methyl 4-methylsalicylate, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methyl 4-methylsalicylate, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H64793.06
also known as H64793-06
Price (USD)/ Each
50.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
50.00
Add to cart
Methyl 4-methylsalicylate, 98+%, Thermo Scientific Chemicals
Catalog numberH64793.06
Price (USD)/ Each
50.00
-
Add to cart
Chemical Identifiers
CAS4670-56-8
IUPAC Namemethyl 2-hydroxy-4-methylbenzoate
Molecular FormulaC9H10O3
InChI KeyUITFCFWKYAOJEJ-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC=C(C)C=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>98.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only