Thermo Scientific Chemicals

Pyrazine-2,5-dicarboxylic acid, 95%, Thermo Scientific Chemicals

Catalog number: H64795.03
1 g, Each
Thermo Scientific Chemicals

Pyrazine-2,5-dicarboxylic acid, 95%, Thermo Scientific Chemicals

Catalog number: H64795.03
1 g, Each
Quantity
Catalog number: H64795.03
also known as H64795-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
122-05-4
IUPAC Name
pyrazine-2,5-dicarboxylic acid
Molecular Formula
C6H4N2O4
InChI Key
GMIOYJQLNFNGPR-UHFFFAOYSA-N
SMILES
OC(=O)C1=CN=C(C=N1)C(O)=O
Assay (HPLC)
>94.0%

Description

It is an pyrazine used for proteomics research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an pyrazine used for proteomics research.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text