Methyl 2-chloropyridine-3-carboxylate, 98%, Thermo Scientific Chemicals
Methyl 2-chloropyridine-3-carboxylate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methyl 2-chloropyridine-3-carboxylate, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Catalog number H64798.22
also known as H64798-22
Price (USD)/ Each
273.00
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
273.00
Add to cart
Methyl 2-chloropyridine-3-carboxylate, 98%, Thermo Scientific Chemicals
Catalog numberH64798.22
Price (USD)/ Each
273.00
-
Add to cart
Chemical Identifiers
CAS40134-18-7
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only