1-Bromo-5-fluoropentane was used to prepare 18F-labeled (anilido)piperidine derivatives as ?-opioid receptor imaging agents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Bromo-5-fluoropentane was used to prepare 18F-labeled (anilido)piperidine derivatives as μ-opioid receptor imaging agents.
Solubility
Soluble in methanol (in methanol). Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
RUO – Research Use Only
General References:
- Friedrich W. Hoffmann. Aliphatic Fluorides. Ii. 1-Halogeno-Ι-Fluoroalkanes1. J. Org. Chem.1950 15(2), 221-224.
- J. Včelák; M. G. Voronkov; V. B. Puchnarevich; S. V. Kirpichenko; and V. Chvalovský . The exchange of fluorine in (γ-fluoropropyl)trialkylsilanes and n-heptyl fluoride for halogens of Grignard reagents. Collect. Czech. Chem. Commun.1976 41(1), 131-139.