Thermo Scientific Chemicals

8-Bromo-1-octene, 97%, Thermo Scientific Chemicals

Catalog number: H64830.06
5 g, Each
Thermo Scientific Chemicals

8-Bromo-1-octene, 97%, Thermo Scientific Chemicals

Catalog number: H64830.06
5 g, Each
Quantity
Catalog number: H64830.06
also known as H64830-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2695-48-9
IUPAC Name
8-bromooct-1-ene
Molecular Formula
C8H15Br
InChI Key
SNMOMUYLFLGQQS-UHFFFAOYSA-N
SMILES
BrCCCCCCC=C
Assay (GC)
>96.0%

Description

8-Bromo-1-octene has been used in preparation of polymerizable ligand, required for the synthesis of quantum dot-labeled polymer beads. Grignard reagent derived from 8-bromo-1-octene has been used in the synthesis of (2S,3S,5R)-5-[(1R)-1-hydroxy-9-decenyl]-2-pentyltetrahydro-3-furanol. It has been used as building block in natural product synthesis. Also used for the synthesis of more complex pharmaceutical compounds, such as Dithienogermole (DTG) derivatives with varying alkyl chain lengths and pendant functionalities.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
8-Bromo-1-octene has been used in preparation of polymerizable ligand, required for the synthesis of quantum dot-labeled polymer beads. Grignard reagent derived from 8-bromo-1-octene has been used in the synthesis of (2S,3S,5R)-5-[(1R)-1-hydroxy-9-decenyl]-2-pentyltetrahydro-3-furanol. It has been used as building block in natural product synthesis. Also used for the synthesis of more complex pharmaceutical compounds, such as Dithienogermole (DTG) derivatives with varying alkyl chain lengths and pendant functionalities.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text