Thermo Scientific Chemicals

4-Chloro-2-(trifluoromethyl)pyridine, 98%, Thermo Scientific Chemicals

Catalog number: H64833.03
1 g, Each
Thermo Scientific Chemicals

4-Chloro-2-(trifluoromethyl)pyridine, 98%, Thermo Scientific Chemicals

Catalog number: H64833.03
1 g, Each
Quantity
Catalog number: H64833.03
also known as H64833-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
131748-14-6
IUPAC Name
4-chloro-2-(trifluoromethyl)pyridine
Molecular Formula
C6H3ClF3N
InChI Key
FZINIBTZNSPWQR-UHFFFAOYSA-N
SMILES
FC(F)(F)C1=NC=CC(Cl)=C1
Form
Liquid
Appearance (Color)
Colorless
Assay (unspecified)
>97.5%

Description

Chloro(trifluoromethyl)pyridines derivatives are used as model substrates for regioexhaustive functionalization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chloro(trifluoromethyl)pyridines derivatives are used as model substrates for regioexhaustive functionalization.

Solubility
Slightly soluble in water.

Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text