Thermo Scientific Chemicals

2-Amino-4-hydroxypyridine, 98%, Thermo Scientific Chemicals

Catalog number: H64836.03
1 g, Each
Thermo Scientific Chemicals

2-Amino-4-hydroxypyridine, 98%, Thermo Scientific Chemicals

Catalog number: H64836.03
1 g, Each
Quantity
Catalog number: H64836.03
also known as H64836-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
33631-05-9
IUPAC Name
2-amino-1,4-dihydropyridin-4-one
Molecular Formula
C5H6N2O
InChI Key
HQNIMNQVKVPZES-UHFFFAOYSA-N
SMILES
NC1=CC(=O)C=CN1
Assay (HPLC)
>97.5%

Description

Employed as an important intermediate for raw material for organic synthesis, agrochemical, pharmaceutical and dyestuff field

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Employed as an important intermediate for raw material for organic synthesis, agrochemical, pharmaceutical and dyestuff field

Solubility
Slightly soluble in water.

Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text