Thermo Scientific Chemicals

N,N-Dimethylpropionamide, ≥98%, Thermo Scientific Chemicals

Catalog number: H64850.14
25 g, Each
Thermo Scientific Chemicals

N,N-Dimethylpropionamide, ≥98%, Thermo Scientific Chemicals

Catalog number: H64850.14
25 g, Each
Quantity
Catalog number: H64850.14
also known as H64850-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
758-96-3
IUPAC Name
N,N-dimethylpropanamide
Molecular Formula
C5H11NO
InChI Key
MBHINSULENHCMF-UHFFFAOYSA-N
SMILES
CCC(=O)N(C)C
Assay (HPLC)
>98.0%

Description

N,N-Dimethylpropionamide has been used in the preparation of AST-type zeolite with a plate morphology. N,N-Dimethylpropionamide is strong donor solvent and coordinate to metal ions through the carbonyl oxygen atom.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Dimethylpropionamide has been used in the preparation of AST-type zeolite with a plate morphology. N,N-Dimethylpropionamide is strong donor solvent and coordinate to metal ions through the carbonyl oxygen atom.

Solubility
Soluble in water.

Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text