Thermo Scientific Chemicals

3-Aminopyridazine, 97%, Thermo Scientific Chemicals

Catalog number: H64859.06
5 g, Each
Thermo Scientific Chemicals

3-Aminopyridazine, 97%, Thermo Scientific Chemicals

Catalog number: H64859.06
5 g, Each
Quantity
Catalog number: H64859.06
also known as H64859-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5469-70-5
IUPAC Name
pyridazin-3-amine
Molecular Formula
C4H5N3
InChI Key
LETVJWLLIMJADE-UHFFFAOYSA-N
SMILES
NC1=CC=CN=N1
Appearance (Color)
White to pale yellow
Form
Solid
Assay (HPLC)
>96.0%

Description

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text