Employed as an important intermediate for syntheses material and pharmaceutical.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Employed as an important intermediate for syntheses material and pharmaceutical.
Solubility
Soluble in acetone, ethanol and methanol.
Notes
Store in freezer at (-20°C). Keep container tightly closed. Store away from oxidizing agent.
RUO – Research Use Only
Maria E de Mesquita; Gilberto F de Sá; Oscar L Malta. Spectroscopic studies of the Eu(III) and Gd(III) tris(3-aminopyridine-2-carboxylic acid) complexes. Journal of Alloys and Compounds.1997 250(1-2), 417-421.
Gerald E. Dunn; Harald F. Thimm; Rajani K. Mohanty. Kinetics and mechanism of decarboxylation of some pyridinecarboxylic acids in aqueous solution. III. 3-Hydroxy- and 3-aminopyridine-2-carboxylic acids. Canadian Journal of Chemistry.1979 57(9), 1098-1104.