Thermo Scientific Chemicals

2-Bromo-4'-methylpropiophenone, 98%, Thermo Scientific Chemicals

Catalog number: H64881.06
5 g, Each
Thermo Scientific Chemicals

2-Bromo-4'-methylpropiophenone, 98%, Thermo Scientific Chemicals

Catalog number: H64881.06
5 g, Each
Quantity
Catalog number: H64881.06
also known as H64881-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1451-82-7
IUPAC Name
2-bromo-1-(4-methylphenyl)propan-1-one
Molecular Formula
C10H11BrO
InChI Key
OZLUPIIIHOOPNQ-UHFFFAOYNA-N
SMILES
CC(Br)C(=O)C1=CC=C(C)C=C1
Assay (HPLC)
>97.5%

Description

It employed as an important intermediate for raw material for organic synthesis, agrochemical, pharmaceutical and dyestuff field. Also used as intermediate for 4-methylmethcathinone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It employed as an important intermediate for raw material for organic synthesis, agrochemical, pharmaceutical and dyestuff field. Also used as intermediate for 4-methylmethcathinone.

Solubility
Soluble in chloroform and methanol.

Notes
Store at 4°C. Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text