Thermo Scientific Chemicals

2-Bromo-5-hydroxybenzaldehyde, 95%, Thermo Scientific Chemicals

Catalog number: H64884.06
5 g, Each
Thermo Scientific Chemicals

2-Bromo-5-hydroxybenzaldehyde, 95%, Thermo Scientific Chemicals

Catalog number: H64884.06
5 g, Each
Quantity
Catalog number: H64884.06
also known as H64884-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2973-80-0
Assay (HPLC)
>94.0%

Description

2-Bromo-5-hydroxybenzaldehyde is a useful reactant used in the synthesis of PDE4 inhibitors, BCL-XL and cancel cell growth inhibitors. Also an reactant used in the preparation of many pharmaceutical agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-5-hydroxybenzaldehyde is a useful reactant used in the synthesis of PDE4 inhibitors, BCL-XL and cancel cell growth inhibitors. Also an reactant used in the preparation of many pharmaceutical agents.

Solubility
Soluble in chloroform, dichloromethane and ethyl acetate.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text