Thermo Scientific Chemicals

4-(Diphenylamino)benzaldehyde, 98%, Thermo Scientific Chemicals

Catalog number: H64887.03
1 g, Each
Thermo Scientific Chemicals

4-(Diphenylamino)benzaldehyde, 98%, Thermo Scientific Chemicals

Catalog number: H64887.03
1 g, Each
Quantity
Catalog number: H64887.03
also known as H64887-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4181-05-9
IUPAC Name
4-(diphenylamino)benzaldehyde
Molecular Formula
C19H15NO
InChI Key
UESSERYYFWCTBU-UHFFFAOYSA-N
SMILES
O=CC1=CC=C(C=C1)N(C1=CC=CC=C1)C1=CC=CC=C1
Appearance (Color)
Cream to yellow
Form
Powder or crystals or crystalline powder
Infrared spectrum
Conforms
Purity
≥97.5%

Description

It is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediate.

Solubility
Soluble in methanol.

Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text