Thermo Scientific Chemicals

2,5-Dihydroxyterephthalic acid, 97%, Thermo Scientific Chemicals

Catalog number: H64888.06
5 g, Each
Thermo Scientific Chemicals

2,5-Dihydroxyterephthalic acid, 97%, Thermo Scientific Chemicals

Catalog number: H64888.06
5 g, Each
Quantity
Catalog number: H64888.06
also known as H64888-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
610-92-4
IUPAC Name
2,5-dihydroxybenzene-1,4-dicarboxylic acid
Molecular Formula
C8H6O6
InChI Key
OYFRNYNHAZOYNF-UHFFFAOYSA-N
SMILES
OC(=O)C1=CC(O)=C(C=C1O)C(O)=O
Assay (HPLC)
?96.0%
Appearance (Color)
Yellow
Identification (FTIR)
conforms
Form
Powder

Description

Employed as an important intermediate for raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Employed as an important intermediate for raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

Solubility
Soluble in hot dimethylformamide.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text